Human Recombinant LAMa5 (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPC078HU01
MSPP-RPC078HU1EA 480 USD
MSPP-RPC078HU1
Human Recombinant LAMa5 (prokaryotic)(from E. coli)
Proteins and Peptides
This is a LAMa5 recombinant protein (prokaryotic), Human is sequencing from Pro2736~Cys2929 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. Laminin subunit alpha-5 belongs to the alpha subfamily of laminin chains and is a major component of basement membranes.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.