Mouse Recombinant Hepc (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB979MU01
MSPP-RPB979MU1EA 400 USD
MSPP-RPB979MU1
Mouse Recombinant Hepc (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a Hepc recombinant protein (prokaryotic), Mouse is sequencing from Thr24~Thr83 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Hepcidin is a peptide hormone produced by the liver discovered in 2000, that appears to be the master regulator of iron homeostasis in humans and other mammals. In humans, HAMP is the gene that encodes for hepcidin. Hepcidin preprohormone, prohormone and hormone size are 84, 60 and 25 amino acids respectively. 20 and 22 amino acid forms of Hepcidin also exist in the urine. The N terminal region is required for function, and deletion of 5 N-terminal peptides results in a loss of function. Hepcidin has a beta-sheet rich structure with 4 disulfide bonds. Hepcidin directly inhibits ferroportin, a protein that transports iron out of cells that store it. This prevents enterocytes of the intestines from secreting iron into the hepatic portal system, reducing iron absorption.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.