Human Recombinant GAPDH (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB932HU02
MSPP-RPB932HU2EA 498 USD
MSPP-RPB932HU2
Human Recombinant GAPDH (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a GAPDH recombinant protein (prokaryotic), Human is sequencing from Thr154~Val324 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Glyceraldehyde 3-phosphate dehydrogenase is an enzyme that catalyzes the sixth step of glycolysis and thus serves to break down glucose for energy and carbon molecules. In addition to this long established metabolic function, GAPDH has recently been implicated in several non-metabolic processes, including transcription activation, initiation of apoptosis , and ER to Golgi vesicle shuttling. Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) catalyses the conversion of glyceraldehyde 3-phosphate as the name indicates. This is the 6th step of the breakDaown of glucose (glycolysis), an important pathway of energy and carbon molecule supply located in the cytosol of eukaryotic cells. Glyceraldehyde 3-phosphate is converted to D-glycerate 1,3-bisphosphate in two coupled steps.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.