Human Recombinant NFkB (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB824HU01
MSPP-RPB824HU1EA 350 USD
MSPP-RPB824HU1
Human Recombinant NFkB (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a NFkB recombinant protein (prokaryotic), Human is sequencing from Pro42~Met367 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Nuclear factor NF-kappa-B p105 subunit isa 105 kDa protein which can undergo cotranslational processing by the 26S proteasome to produce a 50 kDa protein. The 105 kDa protein is a Rel protein-specific transcription inhibitor and the 50 kDa protein is a DNA binding subunit of the NF-kappaB (NF-κB) protein complex.
NF-κB is a transcription factor that is activated by various intra- and extra-cellular stimuli such as cytokines, oxidant-free radicals, ultraviolet irradiation, and bacterial or viral products. Activated NF-κB translocates into the nucleus and stimulates the expression of genes involved in a wide variety of biological functions; over 200 known genes are targets of NF-κB in various cell types, under specific conditions.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.