Human Recombinant S100A9 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB793HU01
MSPP-RPB793HU1EA 320 USD
MSPP-RPB793HU1
Human Recombinant S100A9 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a S100A9 recombinant protein (prokaryotic), Human is sequencing from Met1~Pro114 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
S100 calcium binding protein A9 has been implicated in the abnormal differentiation of myeloid cells in the stroma of cancer. This contributes to creating an overall immunosuppressive microenvironemnt that may contribute to the inability of a protective or therapeutic cellular immune response to be generated by the tumor-bearing host. Outside of malignancy, S100A9 in association with its dimerization partner, S100A8 (MRP8 or calgranulin A) signals for lymphocyte recruitment in sites of inflammation. S100A9/A8 (synonyma: Calgranulin A/B; Calprotectin) are also regarded as marker proteins for a number of inflammatory diseases in humans, especially in rheumatoid arthritis.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.