Rat Recombinant TSLP (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB320RA01
MSPP-RPB320RA1EA 420 USD
MSPP-RPB320RA1
Rat Recombinant TSLP (prokaryotic)(from E. coli)
Proteins and Peptides
This is a TSLP recombinant protein (prokaryotic), Rat is sequencing from Leu75~Ser196 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
The cytokine thymic stromal lymphopoietin (TSLP) drives immature B cell development in vitro and may regulate T helper type 2 responses. Thymic stromal lymphopoietin, also known as TSLP is a human gene.This gene encodes a hemopoietic cytokine proposed to signal through a heterodimeric receptor complex composed of the thymic stromal lymphopoietin receptor and the IL-7R alpha chain. It mainly impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c dendritic cells. Alternative splicing of this gene results in two transcript variants. TSLP is an interleukin (IL)-7-like cytokine produced by epithelial cells and triggers dendritic cell-mediated Th2 type allergic inflammatory responses.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.