Human Recombinant ERa (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPB050HU01
MSPP-RPB050HU1EA 450 USD
MSPP-RPB050HU1
Human Recombinant ERa (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a ERa recombinant protein (prokaryotic), Human is sequencing from Ser178~Met438 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Estrogen receptor structure-function is a vast topic and the subject of very active current research. Nuclear receptors are a large family of structurally related ligand-inducible transcription factors, including steroid receptors (SRs), thyroid/retinoids receptors (TR, RARs and RXRs), vitamin D receptors (VDR), LXR, PPARs, estrogen receptors (ERα and ERβ), and orphan receptors for which no ligand has been yet identified. While having in common a modular structure, they are activated by distinct lipophilic small molecules such as glucocorticoids, progesterone, estrogens, retinoids, and fatty acid derivatives.All nuclear receptors have a hydrophobic pocket into which its specific ligand binds, with helix 12 (H12) being the key response element of NRs.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.