Human Recombinant CHRNa1 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA981HU01
MSPP-RPA981HU1EA 406 USD
MSPP-RPA981HU1
Human Recombinant CHRNa1 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a CHRNa1 recombinant protein (prokaryotic), Human is sequencing from Ser21~Leu255 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
nAChRs, are cholinergic receptors that form ligand-gated ion channels in the plasma membranes of certain neurons. Being ionotropic receptors, nAChRs are directly linked to an ion channel and do not make use of a second messenger as metabotropic receptors do.The muscle acetylcholine receptor consiststs of 5 subunits of 4 different types: 2 alpha isoforms and 1 each of beta, gamma, and delta subunits.2 This gene encodes an alpha subunit that plays a role in acetlycholine binding/channel gating. Alternatively spliced transcript variants encoding different isoforms have been identified. Keiger et al. (2003) surveyed the developmental expression of nicotinic receptors, including CHRNA1, in chick and human spinal cord.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.