Rat Recombinant Cys-C (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA896RA01
MSPP-RPA896RA1EA 452 USD
MSPP-RPA896RA1
Rat Recombinant Cys-C (prokaryotic)(from E. coli)
Proteins and Peptides
This is a Cys-C recombinant protein (prokaryotic), Rat is sequencing from Gly21~Ala140 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Cystatin C: A serum protein that is filtered out of the blood by the kidneys and that serves as a measure of kidney function. Cystatin C is produced steadily by all types of nucleated cells in the body. Its low molecular mass allows it to be freely filtered by the glomerular membrane in the kidney. Its concentration in blood correlates with the glomerular filtration rate. The levels of cystatin C are independent of weight and height, muscle mass, age (over a year of age), and sex. Measurements can be made and interpreted from a single random sample. Cystatin C is a better marker of the glomerular filtration rate and hence of kidney function than creatinine which was the most commonly used measure of kidney function.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.