Human Recombinant SAA (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPA885HU01
MSPP-RPA885HU1EA 300 USD
MSPP-RPA885HU1
Human Recombinant SAA (prokaryotic) (from E.coli)
Proteins and Peptides
This is a SAA recombinant protein (prokaryotic), Human is sequencing from Arg19~Tyr122 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Serum amyloid A protein is a protein encoded by the SAA1 gene. The serum amyloid A (SAA) protein is an acute phase apolipoprotein reactant produced mainly by hepatocytes and under regulation of inflammatory cytokines (summarized by Lundmark et al. 2002). The SAA cleavage product, designated amyloid protein A (AA), is deposited systemically as amyloid in vital organs including the liver, spleen, and kidneys in patients with chronic inflammatory diseases. There is very limited structural information about SAA because of its inherent poor solubility in the apolipoprotein form. It is intriguing to understand how such a small protein is able to mediate or directly carry out such a wide range of functions related to inflammatory reaction and other hostdefense mechanisms.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.