Human Recombinant CRP (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA821HU01
MSPP-RPA821HU1EA 210 USD
MSPP-RPA821HU1
Human Recombinant CRP (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a CRP recombinant protein (prokaryotic), Human is sequencing from pH e17~Pro224 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
C-reactive protein is a liver-derived pattern recognition molecule that is increased in inflammatory states. It rapidly increases within hours after tissue injury, and it is suggested that it is part of the innate immune system and contributes to host defense. Since cardiovascular disease is at least in part an inflammatory process, CRP has been investigated in the context of arteriosclerosis and subsequent vascular disorders. Based on multiple epidemiological and intervention studies, minor CRP elevation has been shown to be associated with future major cardiovascular risk. CRP is known for several decades as a nonspecific inflammation marker. High CRP levels are detected in human blood during bacterial, viral and other infections, as well as in noninfectious diseases such as rheumatic disorders and malignancies.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.