Mouse Recombinant PTHrP (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPA819MU01
MSPP-RPA819MU1EA 450 USD
MSPP-RPA819MU1
Mouse Recombinant PTHrP (prokaryotic) (from E.coli)
Proteins and Peptides
This is a PTHrP recombinant protein (prokaryotic), Mouse is sequencing from Ala37~His175 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Parathyroid hormone-related protein (or PTHrP) is a protein occasionally secreted by cancer cells (breast cancer, certain types of lung cancer including squamous cell carcinoma). However, it also has normal functions. PTHrP acts as an endocrine, autocrine/paracrine, and intracrine hormone. PTHrP is related in function to the 'normal' parathyroid hormone. When a tumor secretes PTHrP, this can lead to hypercalcemia. As this is sometimes the first sign of the malignancy, hypercalcemia caused by PTHrP is considered a paraneoplastic phenomenon. PTHrP shares the same N-terminal end as parathyroid hormone and therefore it can bind to the same receptor (the Type I PTH receptor). PTHrP aids in normal mammary gland development and lactation as well as placental transfer of calcium.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.