Human Recombinant MBP (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA539HU01
MSPP-RPA539HU1EA 280 USD
MSPP-RPA539HU1
Human Recombinant MBP (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a MBP recombinant protein (prokaryotic), Human is sequencing from Val113~Met183 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Myelin basic protein (MBP) is a protein believed to be important in the process of myelination of nerves in the central nervous system (CNS). Myelin Basic Protein (MBP) is the major constituent of the CNS myelin synthesized by oligodendrocytes and Schwann cells. MBP, PLP (proteolipid proteins) and MOG (myelin oligodendrocyte glycoprotein) are potential primary encephalitogenic target antigens in multiple sclerosis.MBP was initially sequenced in 1979 after isolation from myelin membranes. Since that time, knockout mice deficient in MBP have been developed which showed decreased amounts of CNS myelination and a progressive disorder characterized by tremors, seizures, and early death. The gene for MBP is on chromosome 18; the protein localizes to the CNS and to various cells of the hematopoietic system.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.