Human Recombinant APOA1 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA519HU01
MSPP-RPA519HU1EA 300 USD
MSPP-RPA519HU1
Human Recombinant APOA1 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a APOA1 recombinant protein (prokaryotic), Human is sequencing from Gln122~Gln267 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Apolipoprotein A-I  the major protein component of high density lipoprotein (HDL) in plasma. The protein promotes cholesterol efflux from tissues to the liver for excretion. It is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. ApoA-I was also isolated as a prostacyclin (PGI2) stabilizing factor, and thus may have an anticlotting effect. Defects in the gene encoding it are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis.ApoA-1 Milano is a naturally occurring mutant of ApoA-I, found in a family descended from a single couple of the 18th century.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.