Human Recombinant PLP1 (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPA417HU01
MSPP-RPA417HU1EA 350 USD
MSPP-RPA417HU1
Human Recombinant PLP1 (prokaryotic) (from E.coli)
Proteins and Peptides
This is a PLP1 recombinant protein (prokaryotic), Human is sequencing from Leu129~Phe242 with 80 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Proteolipid protein 1 is a protein associated with Pelizaeus-Merzbacher disease. It is a 4 transmembrane domain protein which binds strongly to other copies of itself on the extracellular side of the membrane. In a myelin sheath, as the layers of myelin wraps come together, PLP will bind itself and tightly hold the cellular membranes together.This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. The encoded protein functions in myelination. This protein may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations associated with this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.