Human Recombinant HNRPA2B1 (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA323HU01
MSPP-RPA323HU1EA 480 USD
MSPP-RPA323HU1
Human Recombinant HNRPA2B1 (prokaryotic)(from E. coli)
Proteins and Peptides
This is a HNRPA2B1 recombinant protein (prokaryotic), Human is sequencing from Arg21~Gly103 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Heterogeneous nuclear ribonucleoproteins A2/B1 is a protein belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.