Mouse Recombinant IL4 (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPA077MU01
MSPP-RPA077MU1EA 450 USD
MSPP-RPA077MU1
Mouse Recombinant IL4 (prokaryotic) (from E.coli)
Proteins and Peptides
This is a IL4 recombinant protein (prokaryotic), Mouse is sequencing from His21~Ser140 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Interleukin 4 is a pleiotropic cytokine produced primarily by activated T lymphocytes,mast cells and basophils. IL-4 has multiple immune response-modulating functions on a variety of cell types. It is an important modulator of the differentiation of precursor T helper cells to the Th2 subset that mediates humoral immunity and modulates antibody production. In addition, IL-4 has also been shown to have anti-tumor activity both in vivoand in vitro. The biological effects of IL-4 are mediated by specific cell surface receptor complexes. One type of functional IL-4 receptor complex consists of the IL-4-binding subunit and a second chain, designated the common c chain because it has also been identified as a component of the receptor complexes for IL-4, IL-7, IL-9 and IL-15.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.