Rat Recombinant GFAP (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPA068RA02
MSPP-RPA068RA2EA 350 USD
MSPP-RPA068RA2
Rat Recombinant GFAP (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a GFAP recombinant protein (prokaryotic), Rat is sequencing from Met1~Met430 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
GFAP is an intermediate filament (IF) protein that is found in glial cells such as astrocytes. It is closely related to its non-epithelial family members, vimentin, desmin, and peripherin, which are all involved in the structure and function of the cell’s cytoskeleton. GFAP helps to maintain astrocyte mechanical strength, as well as the shape of cells.GFAP is expressed in the central nervous system in astrocyte cells. It is involved in many cellular functioning processes, such as cell structure and movement, cell communication, and the functioning of the blood brain barrier. The predicted amino acid sequence indicated that GFAP shares structural similarities particularly in the central rod domain and to a lesser degree in the carboxyl-terminal domain with other IF proteins found in nonepithelial cell types.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.