Human Recombinant IFNg (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPA049HU01
MSPP-RPA049HU1EA 300 USD
MSPP-RPA049HU1
Human Recombinant IFNg (prokaryotic) (from E.coli)
Proteins and Peptides
This is a IFNg recombinant protein (prokaryotic), Human is sequencing from Gln24~Gln166 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Interferon gamma (IFN-gama ) is a multifunctional protein first observed as an antiviral activity in cultures of Sindbis virus-infected leukocytes stimulated by PHA. Produced by T lymphocytes and natural killer (NK) cells, IFN-γ is now known to be both an inhibitor of viral replication and a regulator of numerous immunological functions. IFN-γ influences the class of antibody produced by B cells, upregulates class I and II MHC complex antigens and increases. The efficiency of macrophage-mediated killing of intracellular parasites. Most of the activities attributed to IFN-γ are believed to be mediated by IFN-γ-induced proteins. The appearance of such proteins is a consequence of IFN-γ binding to a specific receptor that is distinct from the receptor for IFN-αandβ.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.