Pencil Column Heater
Supplier: PHOENIX S&T INC MS
PST-CH-20U
MSPP-PSTCH-20UEA 1966.15 USD
MSPP-PSTCH-20U
Pencil Column Heater
Heaters Column Heaters
Pencil column heater improve reproducibility, mass transfer and dramatically reduce run pressures and carry-overs.
  • Designed specifically for nano/microspray applications to increase resolution and throughput
  • Temperature range between ambient and 100 °C, with stability control to 0.2 °C