Membrane Filters, Polycarbonate, Sterlitech
Supplier: STERLITECH MS
PCT3013100
MSPP-PCT301310PK 119.86 USD
MSPP-PCT301310
Membrane Filters, Polycarbonate, Sterlitech
Filters Membrane Filters
Polycarbonate Track Etch (PCTE) membranes are made from a thin, microporous polycarbonate film material.
Ideal for use in blood assays and high-purity and general filtration.