Anti-ND5 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAS952RA01
MSPP-PAS952RA1EA 418 USD
MSPP-PAS952RA1
Anti-ND5 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to NADH Dehydrogenase 5 (ND5), derived from recombinant ND5(Arg424~Pro482), is reactive with Rat/Human/Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The MT-ND5 is part of a large enzyme complex known as complex I, which is active in mitochondria. Mitochondria are structures within cells that convert the energy from food into a form that cells can use. Mutations in the MT-ND5 gene are responsible for a small percentage of all cases of mitochondrial encephalomyopathy, lactic acidosis, and stroke-like episodes. These mutations alter single DNA building blocks (nucleotides) in the gene. A particular mutation has been identified in at least five people with the characteristic features of MELAS; this mutation replaces the nucleotide guanine with the nucleotide adenine at gene position 13513. Most of the MT-ND5 mutations that cause MELAS have been shown to reduce the activity of complex I, which disrupts energy production within mitochondria.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ND5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR