Anti-FOLR4 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAS334MU01
MSPP-PAS334MU1EA 396 USD
MSPP-PAS334MU1
Anti-FOLR4 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Folate Receptor 4 (FOLR4), derived from recombinant FOLR4(Cys56~Pro234), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The mouse gene (Folbp3) was sequenced and found to predict a 28.2 kDa protein that consists of 244 amino acids that is highly expressed in both the thymus and spleen, suggesting a potential role in the immune system.,The human gene (FR-delta) is mapped to chromosome 11q14, and predicts a 27.7 kDa protein that is comprised of 241 amino acids. However, expression of the human gene was not detected in 59 samples from both adult and embryonic tissue sources, suggesting a highly restricted spatial/temporal expression pattern, an alternatively spliced variant or an additional FR pseudogene. Using T31 mouse radiation hybrid mapping, Folbp3 was mapped to a region on mouse chromosome 9 that is syntenic to human chromosome 19p13.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: FOLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR