Anti-HYAL1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAS092HU01
MSPP-PAS092HU1EA 386 USD
MSPP-PAS092HU1
Anti-HYAL1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Hyaluronoglucosaminidase 1 (HYAL1), derived from recombinant HYAL1 (Phe22~Trp435), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Hyaluronan, one of the principal glycosaminoglycans of the extracellular matrix, is a high molecular weight polymer of repeating units of N-acetylglucosamine D-glucuronic acid that is synthesized in the plasma membrane of cells and translocated to the pericellular space. It is believed to have numerous important biologic functions, including modulation of cell proliferation, migration, and differentiation, as well as the regulation of extracellular water and protein homeostasis. The predicted 435-amino acid protein is 40% identical to the sperm-specific hyaluronidase SPAM1. Cells transfected with the HYAL1 cDNA secreted hyaluronidase activity that was biochemically indistinguishable from plasma hyaluronidase. Screening of cDNA libraries by PCR indicated that HYAL1 is expressed in multiple tissues, but not in brain.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: HYAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR