Anti-MOSC1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAR574HU01
MSPP-PAR574HU1EA 386 USD
MSPP-PAR574HU1
Anti-MOSC1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to MOCO Sulphurase C-Terminal Domain Containing Protein 1 (MOSC1), derived from recombinant MOSC1(Arg41~Leu335), is reactive with Human/Mouse/Rat/Bovine.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

MOSC1 has been reported to reduce amidoximes to amidines. These MOSC domains contain an absolutely conserved cysteine and occur either as stand-alone forms such as the bacterial YiiM proteins, or fused to other domains such as a NifS-like catalytic domain in Molybdenum cofactor sulfurase. The MOSC domain is predicted to be a sulfur-carrier domain that receives sulfur abstracted by the pyridoxal phosphate-dependent NifS-like enzymes, on its conserved cysteine, and delivers it for the formation of diverse sulfur-metal clusters. The identification of this domain may clarify the mechanism of biogenesis of various metallo-enzymes including Molybdenum cofactor-containing enzymes that are compromised in human type II xanthinuria.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MOSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR