Anti-S100A15 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAQ981HU01
MSPP-PAQ981HU1EA 384 USD
MSPP-PAQ981HU1
Anti-S100A15 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to S100 Calcium Binding Protein A15 (S100A15), derived from recombinant S100A15(Ser2~Gln101), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The human calcium-binding protein (hS100A15) was first identified in inflamed hyperplastic psoriatic skin, where the S100A15 gene is transcribed into two mRNA splice variants, hS100A15-S and hS100A15-L. In cultured keratinocytes, IL-1beta and Th1 cytokines significantly induced hS100A15-L compared with hS100A15-S. In contrast, Th2-derived cytokines had no influence on the expression of either hS100A15 splice variant. Differentiation of human keratinocytes induced by 1.2 mm calcium resulted in the upregulation of both hS100A15 mRNA isoforms. Overexpression of hS100A15 in chronic inflammatory skin diseases and regulation by inflammatory cytokines and calcium suggest that hS100A15 is involved in Th1-associated epithelial responses and epidermal maturation in normal and diseased human skin.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: S100A15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR