Anti-FAM19A3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAQ975MU01
MSPP-PAQ975MU1EA 396 USD
MSPP-PAQ975MU1
Anti-FAM19A3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Family With Sequence Similarity 19, Member A3 (FAM19A3), derived from recombinant FAM19A3 (Ser30~Lys129), is reactive with Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

FAM19A3 is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA genes encode proteins of approximately 100 amino acids that contain conserved cysteine residues at fixed positions. TAFA-1 to -4 are more closely related to each other than to TAFA-5, in which a conserved motif including CC in TAFA-1 to -4 is not present. In H. sapiens, TAFA-3 has two isoforms formed by alternative splicing. Sequence homology analyses reveal that TAFA proteins appear distantly related to MIP-1alpha, a member of the CC-chemokine family. TAFA mRNAs are highly expressed in specific brain regions, with little expression seen in other tissues.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: FAM19A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR