Anti-PI16 Rabbit Polyclonal Antibody

Supplier: 0000027876
PAQ943MU01
MSPP-PAQ943MU1EA 396 USD
MSPP-PAQ943MU1
Anti-PI16 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Peptidase Inhibitor 16 (PI16), derived from recombinant PI16 (Leu21~Lys468), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

PI16 is a putative serine protease inhibitor. PI16 may serve as a marker following prostatectomy for prostate cancer. This protein binds with high affinity to PSP94., PI16 expression is enhanced after serum stimulation of primary cardiomyocytes and is rapidly secreted into the conditioned medium of cardiomyocytes. PI16 is overexpressed in the hypertrophic and failing heart and inhibits cardiomyocyte growth. Suppression of endogenous PI16 expression in primary cardiomyocytes significantly enhances cardiomyocyte size. Transgenic mice overexpressing PI16 in a cardiomyocyte-specific manner show normal cardiac function but have smaller hearts with hypotrophic cardiomyocytes.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PI16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR