Anti-DEFb126 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAQ483HU01
MSPP-PAQ483HU1EA 386 USD
MSPP-PAQ483HU1
Anti-DEFb126 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Defensin Beta 126 (DEFb126), derived from recombinant DEFb126(Asn21~Gly111), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

DEFb126 belongs to the beta-defensin family. High-level and epididymis-specific expression. The innate immune system includes antimicrobial peptides that protect multicellular organisms from a diverse spectrum of microorganisms. beta-Defensins comprise one important family of mammalian antimicrobial peptides. Defensins are cysteine-rich cationic polypeptides that are important in the immunologic response to invading microorganisms. DEFb126 is secreted and is a member of the beta defensin protein family. Beta defensin genes are found in several clusters throughout the genome, with this gene mapping to a cluster at 20p13. The encoded protein is highly similar to an epididymal-specific secretory protein (ESP13.2) from cynomolgus monkey.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: DEFb126
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR