Anti-PABPC1L Rabbit Polyclonal Antibody

Supplier: 0000027876
PAQ477HU01
MSPP-PAQ477HU1EA 386 USD
MSPP-PAQ477HU1
Anti-PABPC1L Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Poly A Binding Protein Cytoplasmic 1 Like Protein (PABPC1L), derived from recombinant PABPC1L(Ser514~Pro609), is reactive with Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

PABPC1L belongs to the polyadenylate-binding protein type-1 family. Recent linkage studies and association analyses indicate the presence of at least one type 2 diabetes susceptibility gene in human chromosome region 20q12-q13.1. The map currently provides near-continuous coverage between SGC32867 and WI-17676. EST database searches and genomic sequence alignments of ESTs, mRNAs, and UniGene clusters enabled the annotation of the sequence interval with experimentally confirmed and putative transcripts. The combination of the BAC transcript map, YAC-to-BAC scaffold, and reference Human Genome Project sequence provides a powerful integrated resource for future genomic analysis of this region.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PABPC1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR