Anti-MXRA5 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAP859HU01
MSPP-PAP859HU1EA 386 USD
MSPP-PAP859HU1
Anti-MXRA5 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Matrix Remodelling Associated Protein 5 (MXRA5), derived from recombinant MXRA5 (Pro2542~Phe2828), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Adlican is one of the matrix-remodelling associated proteins. This protein contains 7 leucine-rich repeats and 12 immunoglobulin-like C2-type domains related to perlecan. This gene has a pseudogene on chromosome Y., In order to discover global gene expression patterns characterizing subgroups of colon cancer, microarrays were hybridized to labeled RNAs obtained from seventeen colonic specimens. Using a hierarchical agglomerative method, the samples grouped naturally into two major clusters, in perfect concordance with pathological reports. Using a variant of the unpaired t-test, selected genes were ordered according to an index of importance. RT-PCR was performed on the gro1, B-factor, adlican, and endothelin converting enzyme-1 genes and confirmed microarray findings.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: MXRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR