Anti-PSMD9 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAP779HU01
MSPP-PAP779HU1EA 384 USD
MSPP-PAP779HU1
Anti-PSMD9 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Proteasome 26S Subunit, Non ATPase 9 (PSMD9), derived from recombinant PSMD9(Ser2~Arg223), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

PSMD9 encodes a non-ATPase subunit of the 19S regulator.Sequence analysis predicted that the 222-amino acid Bridge-1 protein shares 98% amino acid similarity with human PSMD9 over the first 184 amino acids but diverges in the C terminus Bridge-1 contains a PDZ-like domain from amino acids 138 to 178, forming 3 beta sheets and 2 alpha helices. SDS-PAGE analysis showed that Bridge-1 is expressed as a 28-kDa protein, close to the deduced value of 25 kD. Using Bridge-1 cDNA as probe, Northern blot analysis detected a 1.0-kb transcript in all rat and human tissues tested, with highest expression in pancreas, testis, kidney, and liver. Immunocytochemistry assessment demonstrated predominant nuclear localization of Bridge-1, with lower levels in cytoplasm.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PSMD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR