Anti-TOMM70A Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAP574HU01
MSPP-PAP574HU1EA 386 USD
MSPP-PAP574HU1
Anti-TOMM70A Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Translocase Of Outer Mitochondrial Membrane 70A (TOMM70A), derived from recombinant TOMM70A(Leu333~Leu608), is reactive with Human/Mouse/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

The translocase of outer mitochondrial membrane (TOM) complex is a multisubunit complex involved in the recognition, unfolding, and translocation of preproteins into the mitochondria.TOMM70A is 29% identical to the mitochondrial precursor protein import receptor of Neurospora crassa. RT-PCR analysis detected ubiquitous expression of KIAA0719, with relatively weak expression in pancreas, testis, and spleen.the cytosolic chaperones HSP90 and HSP70 dock onto a specialized tetratricopeptide domain in the import receptor TOMM70 at the outer mitochondrial membrane. This interaction served to deliver a set of preproteins to the receptor for subsequent membrane translocation dependent on the HSP90 ATPase. Disruption of the chaperone/TOMM70 recognition inhibited the import of these preproteins into mitochondria.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: TOMM70A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR