Anti-RSPO3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAM173HU01
MSPP-PAM173HU1EA 386 USD
MSPP-PAM173HU1
Anti-RSPO3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to R-Spondin 3 (RSPO3), derived from recombinant RSPO3 (Gln22~His272), is reactive with Human/Rat/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

RSPO3 encodes a member of the thrombospondin type 1 repeat supergene family. In addition, the protein contains a furin-like cysteine-rich region. Furin-like repeat domains have been found in a variety of eukaryotic proteins involved in the mechanism of signal transduction by receptor tyrosine kinases. The 272-amino acid protein has an N-terminal cys-rich domain (CRD) and a central thrombospondin type-1 (THBS1) repeat (TSR) domain. Northern blot analysis detected 3.0- and 4.0-kb transcripts in placenta, lung, and skeletal muscle. RT-PCR revealed ubiquitous expression, with highest levels in placenta, small intestine, fetal thymus, and lymph node. RSPO3 expression was detected in all tumor tissues examined except a lung carcinoma and an ovarian carcinoma.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: RSPO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR