Anti-PRDM1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAM058HU01
MSPP-PAM058HU1EA 386 USD
MSPP-PAM058HU1
Anti-PRDM1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to PR Domain Containing Protein 1 (PRDM1), derived from recombinant PRDM1 (Leu29~Asp149), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

PRDM1 encodes a protein that acts as a repressor of beta-interferon gene expression. The protein binds specifically to the PRDI (positive regulatory domain I element) of the beta-IFN gene promoter. Transcription of this gene increases upon virus induction. Two alternatively spliced transcript variants that encode different isoforms have been reported.PRDIBF1 is the human homolog of the mouse Blimp1 gene, the expression of which drives the terminal differentiation of B lymphocytes. The author also identified a novel stretch of 100 amino acids, which he named the PR domain, that is conserved between PRDIBF1 and RIZ.The deduced 789-amino acid PRDIBF1 protein has a predicted molecular mass of 88 kDa and contains 5 zinc fingers of the TFIIA type near the C terminus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: PRDM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR