Anti-NFE2L2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL947HU01
MSPP-PAL947HU1EA 386 USD
MSPP-PAL947HU1
Anti-NFE2L2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2), derived from recombinant NFE2L2(Ser400~Lys603), is reactive with Human/Pig.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Nrf2 is a master regulator of the antioxidant response. The antioxidant response is important for the amelioration of oxidative stress. Oxidative stress can result in cancer, cardiovascular diseases, inflammation, neurological diseases, and renal disease. Because Nrf2 is able to induce genes important in combating oxidative stress, thereby activating the bodies own protective response, it is able to protect from a variety of [oxidative stress]-related complications, even in situations where the administration of exogenous antioxidants have failed.NFE2, NFE2L1, and NFE2L2 comprise a family of human genes encoding basic leucine zipper (bZIP) transcription factors. They share highly conserved regions that are distinct from other bZIP families, such as JUN and FOS, although remaining regions have diverged considerably from each other.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NFE2L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR