Anti-SEMA5B Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL926HU01
MSPP-PAL926HU1EA 386 USD
MSPP-PAL926HU1
Anti-SEMA5B Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Semaphorin 5B (SEMA5B), derived from recombinant SEMA5B (Leu36~Glu161), is reactive with Human/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Members of the semaphorin protein family, such as SEMA5B, are involved in axonal guidance during neural development. The deduced 1,093-amino acid protein contains an N-terminal signal sequence, followed by a semaphorin domain, 7 thrombospondin type-1 repeats, a putative transmembrane domain, and a short cytoplasmic tail. It also has 13 conserved cysteines and a conserved N-glycosylation site. Northern blot analysis of whole mouse embryos detected Semg at embryonic day 10, and its expression decreased toward birth. During development, Semg was expressed exclusively in the neuroepithelium along the entire anterioposterior axis. In adult animals, Semg was detected only in brain. The deduced 1,202-amino acid protein shares 93% identity with mouse Semg.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: SEMA5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR