Anti-SEMA5A Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL924HU01
MSPP-PAL924HU1EA 386 USD
MSPP-PAL924HU1
Anti-SEMA5A Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Semaphorin 5A (SEMA5A), derived from recombinant SEMA5A (Val319~Gln668), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

SEMA5A belongs to the semaphorin gene family that encodes membrane proteins containing a semaphorin domain and several thrombospondin type-1 repeats. Members of this family are involved in axonal guidance during neural development. This gene has been implicated as an autism susceptibility gene. The deduced 1077-amino acid protein contains an N-terminal signal sequence, followed by a semaphorin domain, 7 thrombospondin type-1 repeats, a putative transmembrane domain, and a short cytoplasmic tail. It also has 13 conserved cysteines and a conserved N-glycosylation site. Northern blot analysis of whole mouse embryos detected 2 Semf transcripts at embryonic day 10, and their expression decreased toward birth.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: SEMA5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR