Anti-SEMA3C Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL919HU01
MSPP-PAL919HU1EA 386 USD
MSPP-PAL919HU1
Anti-SEMA3C Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Semaphorin 3C (SEMA3C), derived from recombinant SEMA3C (Arg28~Asp150), is reactive with Human/Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The 616-amino acid human semaphorin E contains an N-terminal signal sequence of 15 amino acids that is followed by a semaphorin domain. in vitro translation of the semaphorin E cDNA yielded a 70-KD protein. The authors showed that semaphorin E is secreted by mammalian cells. By Northern blot analysis, semaphorin E was expressed as a 5.2-Kb transcript in all human tissues tested. CDDP-sensitive cells transfected with semaphorin E cDNA acquired CDDP-resistance. Semaphorin E was overexpressed in CDDP-resistant cell lines and was induced by diverse chemotherapeutic drugs and by x-ray and UV irradiation. Aberrant semaphorin E protein expression was detected immunohistochemically.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: SEMA3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR