Anti-MIB2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL728MU01
MSPP-PAL728MU1EA 396 USD
MSPP-PAL728MU1
Anti-MIB2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Mindbomb Homolog 2 (MIB2), derived from recombinant MIB2 (Val546~Asn784), is reactive with Mouse.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Skeletrophin was expressed in 23 of 25 benign nevi, but not in 18 of 38 cases of primary skin melanoma. Treatment with a demethylating agent restored skeletrophin expression in a human melanoma cell line.The deduced 1,013-amino acid protein contains a dystrophin (DMD)-like cysteine-rich zinc-binding domain in its N-terminal half and 5 ankyrin repeats in its C-terminal half. Northern blot analysis detected a 3.2-kb transcript that was highly expressed in skeletal muscle and more weakly expressed in brain, heart, and kidney. Western blot analysis detected a 70-kD protein in skeletal muscle, heart, and brain, but not in liver, thymus, or lung. Immunohistochemical analysis of skeletal muscle detected cytoplasmic foci of skeletrophin expression near the cell surface.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: MIB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR