Anti-KEAP1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAL648HU01
MSPP-PAL648HU1EA 384 USD
MSPP-PAL648HU1
Anti-KEAP1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Kelch Like ECH Associated Protein 1 (KEAP1), derived from recombinant KEAP1(Leu327~Cys624), is reactive with Human/Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

KEAP1 encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.Kelch motifs of Keap1 interact with the Neh2 domain of Nrf2 and that this interaction is required for the repression of Nrf2 by Keap1. Fluorescence microscopy demonstrated that Keap1 is expressed in the cytoplasm with Nrf2.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: KEAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR