Anti-Mer Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAK946GE01
MSPP-PAK946GE1EA 550 USD
MSPP-PAK946GE1
Anti-Mer Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Meropenem (Mer), derived from OVA conjugated Mer, is reactive with General species.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

Meropenem is an ultra-broad-spectrum injectable antibiotic used to treat a wide variety of infections. It is a beta-lactam and belongs to the subgroup of carbapenem, similar to imipenem and ertapenem. Meropenem is bactericidal except against Listeria monocytogenes, where it is bacteriostatic. It inhibits bacterial wall synthesis like other beta-lactam antibiotics. In contrast to other beta-lactams, it is highly resistant to degradation by beta-lactamases or cephalosporinases. In general, resistance arises due to mutations in penicillin-binding proteins, production of metallo-beta-lactamases, or resistance to diffusion across the bacterial outer membrane. Unlike imipenem, it is stable to dehydropeptidase-1 and can therefore be given without cilastatin. Meropenem must be administered intravenously. It is supplied as a white crystalline powder to be dissolved in 5% monobasic potassium phosphate solution.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: Mer
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR