Anti-GKN3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAK528MU01
MSPP-PAK528MU1EA 360 USD
MSPP-PAK528MU1
Anti-GKN3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Gastrokine 3 (GKN3), derived from recombinant GKN3 (Tyr38~Leu177), is reactive with Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

A novel gastrokine, Gkn3, marks gastric atrophy and shows evidence of adaptive gene loss in humans. Gkn3 encodes a secreted protein that is co-expressed with trefoil factor (Tff)2 in the distal stomach and discriminates a Griffinia simplicifolia lectin (GS)-II-positive mucus neck cell (MNC) subpopulation in the proximal stomach. In humans, widespread homozygosity for a premature stop codon polymorphism, W59X, has likely rendered GKN3 non-functional. Mouse Gkn3 expression is strongly up-regulated in (Tff2-deficient) gastric atrophy, a pre-cancerous state that is typically associated with Helicobacter pylori and marks a non-proliferative, GS-II positive lineage with features of spasmolytic polypeptide-expressing metaplasia (SPEM). Gkn3 overexpression inhibits proliferation in gastric epithelial cell lines, independently of incubation with recombinant human TFF2 or apoptosis.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please properly store each component based on the instruction.

Type: Primary
Antigen: GKN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR