Anti-NLRP3 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAK115MU01
MSPP-PAK115MU1EA 360 USD
MSPP-PAK115MU1
Anti-NLRP3 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to NLR Family, Pyrin Domain Containing Protein 3 (NLRP3), derived from recombinant NLRP3(Cys671~Trp1033), is reactive with Human/Mouse.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

NLRP3 encodes a pyrin-like protein which contains a pyrin domain, a nucleotide-binding site (NBS) domain, and a leucine-rich repeat (LRR) motif. This protein interacts with apoptosis-associated speck-like protein containing a CARD. Proteins which contain the caspase recruitment domain, CARD, have been shown to be involved in inflammation and immune response. This protein may function as an activator of NF-κB signaling. The protein may play a role in the regulation of inflammation and apoptosis. Mutations in this gene have been associated with a spectrum of dominantly inherited autoinflammatory diseases called cryopyrin-associated periodic syndrome (CAPS). Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NLRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR