Anti-FADD Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAK078MU01
MSPP-PAK078MU1EA 360 USD
MSPP-PAK078MU1
Anti-FADD Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Fas Associating Death Domain Containing Protein (FADD), derived from recombinant FADD (Leu7~Glu178), is reactive with Mouse/Rat.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

FADD is an adaptor molecule that bridges the Fas-receptor, and other death receptors, to caspase-8 through its death domain to form the death inducing signaling complex during apoptosis. The protein encoded by this gene is an adaptor molecule that interacts with various cell surface receptors and mediates cell apoptotic signals. Through its C-terminal death domain, this protein can be recruited by TNFRSF6/Fas-receptor, tumor necrosis factor receptor, TNFRSF25, and TNFSF10/TRAIL-receptor, and thus it participates in the death signaling initiated by these receptors. Interaction of this protein with the receptors unmasks the N-terminal effector domain of this protein, which allows it to recruit caspase-8, and thereby activate the cysteine protease cascade.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR