Anti-NANOG Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAJ992HU01
MSPP-PAJ992HU1EA 350 USD
MSPP-PAJ992HU1
Anti-NANOG Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to NANOG Homeobox Protein (NANOG), derived from recombinant NANOG(Met1~Val305), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

NANOG is a transcription factor critically involved with self-renewal of undifferentiated embryonic stem cells. Human NANOG protein is a 305 amino acid protein with a conserved homeodomain motif that is localized to the nuclear component of cells. The homeodomain region facilitates DNA binding. There are N-terminal, homeodomain, and C-terminal regions in human NANOG protein. Like murine NANOG, N-terminal region of human NANOG is rich in Ser, Thr and Pro residues and C-terminus contains W repeats. The homeodomain in hNANOG ranges from residues 95 to 155. NANOG is a gene expressed in embryonic stem cells (ESCs) and is thought to be a key factor in maintaining pluripotency. NANOG is thought to function in concert with other factors such as POU5F1 and SOX2 to establish ESC identity.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR