Anti-CCDC80 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAJ901HU01
MSPP-PAJ901HU1EA 350 USD
MSPP-PAJ901HU1
Anti-CCDC80 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Coiled Coil Domain Containing Protein 80 (CCDC80), derived from recombinant CCDC80(Pro81~Tyr258), is reactive with Human/Pig.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The Urb protein contains a highly hydrophobic N-terminal signal sequence, indicating that it is a secretory protein, 3 N-glycosylation sites, several pairs of basic amino acids, and a lysine-rich region containing nuclear localization signals. Three consensus repeat domains in Urb share about 30% amino acid identity with the fifth domain of the human SRPX protein, suggesting that Urb may be functionally related to the sushi protein superfamily. The nucleotide sequences of Urb and rat Ssg1 share more than 90% identity, but several mismatches result in a frameshift, and the Ssg1 protein contains only 385 residues. In situ hybridization of mouse brain showed expression of Urb in the anterior olfactory nucleus, dorsal endopiriform nucleus, and choroid plexus; weak expression was observed in the cortex.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: CCDC80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR