Anti-EMILIN2 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAJ161MU01
MSPP-PAJ161MU1EA 360 USD
MSPP-PAJ161MU1
Anti-EMILIN2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Elastin Microfibril Interface Located Protein 2 (EMILIN2), derived from recombinant EMILIN2 (Arg853~Leu1074), is reactive with Mouse/Rat.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

EMILIN2 contains an N-terminal signal peptide with a consensus signal cleavage site, followed by an emilin (EMI) domain, several coiled-coil domains, a proline-rich region, a collagen-like coiled-coil alpha-helical region, and a C-terminal C1q domain. The mature 1023-amino acid protein has a calculated molecular mass of 112 kDa. EMILIN2 also has 8 potential N-glycosylation sites and 20 cysteines, a number of which are clustered as doublets that may form intramolecular disulfide bonds. RNA dot blot analysis detected high EMILIN1 expression in fetal heart and adult lung. Intermediate expression was detected in peripheral leukocytes, placenta, and spinal cord, and low expression was detected in fetal brain, spleen, thymus, and lung, and in adult heart, aorta, testis, bone marrow, small intestine, thymus, lymph node, and appendix.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: EMILIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR