Anti-ITIH1 Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAH779HU01
MSPP-PAH779HU1EA 350 USD
MSPP-PAH779HU1
Anti-ITIH1 Rabbit Polyclonal Antibody
Antibodies
Polyclonal antibody to Inter Alpha-Globulin Inhibitor H1 (ITIH1), derived from recombinant ITIH1(Ser507~Asp819), is reactive with Human.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Salier et al. (1987) characterized cDNAs for the H chain of IATI. Diarra-Mehrpour et al. (1989) showed that human poly(A)-rich RNAs hybrid-selected with various heavy-chain-encoding cDNA clones translate 3 different heavy chains, designated H1 (M(r) 92,000), H2 (ITIH2), and H3 (ITIH3). Two previously characterized heavy-chain cDNA clones were found to correspond to the H1 and H2 chains. The deduced amino acid sequence of the H3 chain was found to be highly similar to those of the H1 (54%) and H2 (44%) chains. Diarra-Mehrpour et al. (1992) used a PCR-based cloning approach and cDNA library screening to isolate the full-length cDNA H1. The results of Northern blot analysis by Bost et al. (1993) indicated that the 2.9-kb ITIH1 mRNA is expressed only in liver.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR