Anti-NEXN Rabbit Polyclonal Antibody

Supplier: CLOUD-CLONE CORP MS
PAH548HU01
MSPP-PAH548HU1EA 350 USD
MSPP-PAH548HU1
Anti-NEXN Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to Nexilin (NEXN), derived from recombinant NEXN (Glu409~Val605), is reactive with Human.

  • High specificity, affinity, and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee

The name nexilin came from the Latin word nexilis, meaning 'bound together.' Full-length b-nexilin contains an N-terminal F-actin-binding domain (ABD), followed by a spacer, a coiled-coil region, a second ABD, and a long C-terminal tail. In comparison, s-nexilin lacks the N-terminal ABD and has an insertion in the C-terminal tail. Immunofluorescence microscopy revealed that both b-nexilin and s-nexilin colocalized with F-actin at focal contacts at the ends of stress fibers., The nexilins did not localize to cell-cell junctions. Western blot analysis detected nexilin proteins with apparent molecular masses above 90 kDa in rat brain, testis, and spleen and in cultured rodent fibroblasts. Neither protein was detected in liver, kidney, or epithelial cell lines.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: NEXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR